Product Details

SNP ID
rs149096836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42374392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTTAATGTGGGCTTTAGTGATA[A/C]ATAAGGAAAGTGGCCTGCTGGGGGT
Phenotype
MIM: 605417 MIM: 174760
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DKK4 PubMed Links

Gene Details

Gene
DKK4
Gene Name
dickkopf WNT signaling pathway inhibitor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014420.2 Intron NP_055235.1
XM_011544488.2 Intron XP_011542790.1
XM_017013316.1 Intron XP_016868805.1
Gene
POLB
Gene Name
DNA polymerase beta
There are no transcripts associated with this gene.

View Full Product Details