Product Details

SNP ID
rs11551243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28538011 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCCCCCCGACTCCTTACCTCCA[G/A]CTCTGTCTCAGCGCCGTGCCCCTCG
Phenotype
MIM: 614812
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
NUPR1 PubMed Links

Gene Details

Gene
NUPR1
Gene Name
nuclear protein 1, transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042483.1 524 UTR 3 NP_001035948.1
NM_012385.2 524 UTR 3 NP_036517.1

View Full Product Details