Product Details

SNP ID
rs17273309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54713337 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTCTGGACCATCATTCGGGAAAC[C/T]GTCTACTGAGAAAGTCGAGAAGGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LILRP2 PubMed Links

Gene Details

Gene
LILRP2
Gene Name
leukocyte immunoglobulin-like receptor pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details