Product Details

SNP ID
rs73669642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128122898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGCTCGTGGCACCGGAGGCTC[A/G]GGGACAGCAGGCCCCGGATGTGCAC
Phenotype
MIM: 608152
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTGES2 PubMed Links

Gene Details

Gene
PTGES2
Gene Name
prostaglandin E synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256335.1 Intron NP_001243264.1
NM_025072.6 Intron NP_079348.1
NM_198938.2 Intron NP_945176.1
XM_005252239.2 Intron XP_005252296.1
XM_011519050.1 Intron XP_011517352.1
XM_011519051.2 Intron XP_011517353.1
Gene
PTGES2-AS1
Gene Name
PTGES2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
SLC25A25-AS1
Gene Name
SLC25A25 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details