Product Details

SNP ID
rs13345394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14880647 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTGTGATGCACAGGTGGAAAATG[C/T]CTTGTACTTCCCCTGAGCTGATGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR7A17 PubMed Links

Gene Details

Gene
OR7A17
Gene Name
olfactory receptor family 7 subfamily A member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030901.1 Intron NP_112163.1

View Full Product Details