Product Details

SNP ID
rs8192583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32195497 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTTGGAGGGCTGGGGGACCACA[A/G]TCAAGTTGAGGTGATCCCCGCTCCG
Phenotype
MIM: 164951 MIM: 176311
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPSM3 PubMed Links

Gene Details

Gene
GPSM3
Gene Name
G-protein signaling modulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276501.1 6091 Intron NP_001263430.1
NM_022107.2 6091 UTR 5 NP_071390.1
Gene
NOTCH4
Gene Name
notch 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004557.3 6091 Silent Mutation GAC,GAT D1984D NP_004548.3
Gene
PBX2
Gene Name
PBX homeobox 2
There are no transcripts associated with this gene.

View Full Product Details