Product Details
- SNP ID
-
rs200033604
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:97757390 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTGTTACCGAGGACCGACTGCGA[C/G]AAAACAGAAAGCGTCCAGCACTGAG
- Phenotype
-
MIM: 615659
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM131
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115906722] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEM131
- Gene Name
- transmembrane protein 131
View Full Product Details