Product Details

SNP ID
rs61745885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56346447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACGTAGCCATCTGTAAACCTCTT[C/T]TGTACGTGATCATCATGGCAGAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR8K1 PubMed Links

Gene Details

Gene
OR8K1
Gene Name
olfactory receptor family 8 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002907.1 409 Silent Mutation CTG,TTG L137L NP_001002907.1

View Full Product Details