Product Details

SNP ID
rs11465293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75813405 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAACAAAGAACATGCAGCAGGGA[A/G]AGGGGATGACCACAGAGCCTAGAAG
Phenotype
MIM: 602495
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL24 PubMed Links

Gene Details

Gene
CCL24
Gene Name
C-C motif chemokine ligand 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002991.2 359 Missense Mutation TCT,TTT S31F NP_002982.2
XM_011516459.2 359 Missense Mutation TCT,TTT S31F XP_011514761.1
XM_011516460.2 359 Missense Mutation TCT,TTT S31F XP_011514762.1

View Full Product Details