Product Details

SNP ID
rs61747888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55603563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGTGTTCCAATAGTATAAAATA[C/T]TGCCACCATCTTGTCCATGGGGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4C11 PubMed Links

Gene Details

Gene
OR4C11
Gene Name
olfactory receptor family 4 subfamily C member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004700.2 836 Missense Mutation ATA,GTA I271V NP_001004700.2

View Full Product Details