Product Details

SNP ID
rs3765005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150659887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAACAAATGAAAGCACCATCATT[C/G]TTAGTTAAATTTCTTCATTTATTAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERICH6 PubMed Links

Gene Details

Gene
ERICH6
Gene Name
glutamate rich 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308234.1 1766 UTR 3 NP_001295163.1
NM_152394.4 1766 UTR 3 NP_689607.2
XM_005247120.3 1766 Intron XP_005247177.1

View Full Product Details