Product Details

SNP ID
rs62000367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:121757331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCATCAGGGTGCTGGGTTTCAGG[A/C]CAGTAAGTTCTTTGTTTTGTATCCA
Phenotype
MIM: 600575
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TSN PubMed Links

Gene Details

Gene
TSN
Gene Name
translin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261401.1 393 Missense Mutation GAC,GCC D53A NP_001248330.1
NM_004622.2 393 Missense Mutation GAC,GCC D53A NP_004613.1

View Full Product Details