Product Details

SNP ID
rs34537598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:56802170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCCAGATAGACACAGGAGCACT[A/G]CTGCTGGCAATGGTCGGGACTGAGG
Phenotype
MIM: 600037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTX2 PubMed Links

Gene Details

Gene
OTX2
Gene Name
orthodenticle homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270523.1 872 Silent Mutation AGC,AGT S145S NP_001257452.1
NM_001270524.1 872 Silent Mutation AGC,AGT S145S NP_001257453.1
NM_001270525.1 872 Silent Mutation AGC,AGT S153S NP_001257454.1
NM_021728.3 872 Silent Mutation AGC,AGT S153S NP_068374.1
NM_172337.2 872 Silent Mutation AGC,AGT S145S NP_758840.1

View Full Product Details