Product Details
- SNP ID
-
hCV25751598
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:55256319 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATAATCATCATTAAAAAGGAACTA[A/G]AAAATTATACATGTTTAAAGTATGT
- Phenotype
-
MIM: 612720
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DHX29
PubMed Links
Gene Details
- Gene
- DHX29
- Gene Name
- DEAH-box helicase 29
There are no transcripts associated with this gene.
View Full Product Details