Product Details

SNP ID
rs35252424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63606358 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGTATGAGGGTCGAGGCCCTGGT[A/G]GCTCTTCTCTGCTCATCGAGGCATT
Phenotype
MIM: 612958
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TACO1 PubMed Links

Gene Details

Gene
TACO1
Gene Name
translational activator of cytochrome c oxidase I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016360.3 645 Missense Mutation AGC,GGC S145G NP_057444.2

View Full Product Details