Product Details

SNP ID
rs112506621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:128965992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTTCTCAGGATTCCTCCCCATC[C/T]CGCTTCGCCCCGGAAAAGCTGACAA
Phenotype
MIM: 604196
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC27A6 PubMed Links

Gene Details

Gene
SLC27A6
Gene Name
solute carrier family 27 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017372.2 690 UTR 5 NP_001017372.1
NM_001317984.1 690 UTR 5 NP_001304913.1
NM_014031.4 690 UTR 5 NP_054750.1

View Full Product Details