Product Details

SNP ID
rs72758134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120859968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTCACAATTGCAGAAGGCCAGG[G/T]ACTCCCCGCCAGAGACTGAGACACA
Phenotype
MIM: 609740
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009936.2 Intron NP_001009936.1
NM_001286840.1 Intron NP_001273769.1
NM_001286842.1 Intron NP_001273771.1
NM_001286843.1 Intron NP_001273772.1
NM_015651.2 Intron NP_056466.1
XM_005251906.2 Intron XP_005251963.1
XM_011518509.2 Intron XP_011516811.1
XM_011518511.2 Intron XP_011516813.1
XM_011518515.2 Intron XP_011516817.1
XM_011518516.2 Intron XP_011516818.1
XM_017014612.1 Intron XP_016870101.1
XM_017014613.1 Intron XP_016870102.1
Gene
PSMD5-AS1
Gene Name
PSMD5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details