Product Details

SNP ID
rs140124846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:37850922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGAATGAATATGATTTTTATCC[C/G]ATTCTTATAACGTCGAGGCTATTTA
Phenotype
MIM: 607421
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NME8 PubMed Links

Gene Details

Gene
NME8
Gene Name
NME/NM23 family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016616.4 Intron NP_057700.3

View Full Product Details