Product Details

SNP ID
rs34803976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93670828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAAGTTCAGATTCTGAAGCAC[G/T]TTCTTCCTCATCAGAGTTTCCTGTT
Phenotype
MIM: 608866 MIM: 600827
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FRA10AC1 PubMed Links

Gene Details

Gene
FRA10AC1
Gene Name
fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145246.4 1119 Missense Mutation NP_660289.2
XM_005269515.4 1119 Missense Mutation XP_005269572.1
XM_005269516.4 1119 Missense Mutation XP_005269573.1
XM_011539259.2 1119 Missense Mutation XP_011537561.1
XM_011539260.2 1119 Missense Mutation XP_011537562.1
XM_017015656.1 1119 Silent Mutation XP_016871145.1
XM_017015657.1 1119 Silent Mutation XP_016871146.1
XM_017015658.1 1119 Silent Mutation XP_016871147.1
XM_017015659.1 1119 Missense Mutation XP_016871148.1
XM_017015660.1 1119 Silent Mutation XP_016871149.1
XM_017015661.1 1119 Missense Mutation XP_016871150.1
XM_017015662.1 1119 UTR 3 XP_016871151.1
Gene
PDE6C
Gene Name
phosphodiesterase 6C
There are no transcripts associated with this gene.

View Full Product Details