Product Details

SNP ID
rs139942301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52832012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCCTCAGAGTGAAGCCAAGGCC[A/G]GCCACCATCATTAAGATGACCCAGG
Phenotype
MIM: 607605
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTA5 PubMed Links

Gene Details

Gene
GSTA5
Gene Name
glutathione S-transferase alpha 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153699.1 Intron NP_714543.1

View Full Product Details