Product Details

SNP ID
rs3826941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44060828 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATGATGGATATAGCAACCCAAAG[A/G]GAAGGGAATTCAGGTAAGAAGCACG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF223 PubMed Links

Gene Details

Gene
ZNF223
Gene Name
zinc finger protein 223
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013361.5 720 Silent Mutation AGA,AGG R74R NP_037493.3
XM_017027258.1 720 Silent Mutation AGA,AGG R74R XP_016882747.1
XM_017027259.1 720 Silent Mutation AGA,AGG R74R XP_016882748.1
XM_017027260.1 720 Intron XP_016882749.1

View Full Product Details