Product Details
- SNP ID
-
rs3751601
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:52112019 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GCGCAGCTGCTTCGGAACGGTTCAC[A/G]GCCGTCTAAGAAGGCCGATTTCTGC
- Phenotype
-
MIM: 606910
MIM: 604447
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BCL2L10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76201546] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BCL2L10
- Gene Name
- BCL2 like 10
- Gene
- GNB5
- Gene Name
- G protein subunit beta 5
There are no transcripts associated with this gene.
View Full Product Details