Product Details

SNP ID
rs3176860
Assay Type
Validated
NCBI dbSNP Submissions
74
Location
Chr.1:100721663 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GTAACCTTAATTGATTCTGCAACAA[A/G]CCGCATGTGTTGCTCAAGCTTCCCA
Phenotype
MIM: 192225
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VCAM1 PubMed Links

Gene Details

Gene
VCAM1
Gene Name
vascular cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001078.3 Intron NP_001069.1
NM_001199834.1 Intron NP_001186763.1
NM_080682.2 Intron NP_542413.1

View Full Product Details