Product Details

SNP ID
rs72908970
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:35184369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGCTAACATCCATAAAAAGATA[A/G]CTTTCTTACTAAAATGCAAGAATTT
Phenotype
MIM: 605199
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SFPQ PubMed Links

Gene Details

Gene
SFPQ
Gene Name
splicing factor proline and glutamine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005066.2 2323 Intron NP_005057.1
XM_005271112.4 2323 UTR 3 XP_005271169.1
XM_005271113.4 2323 UTR 3 XP_005271170.1
XM_005271115.4 2323 Intron XP_005271172.1
XM_011541950.2 2323 UTR 3 XP_011540252.1
XM_017002053.1 2323 UTR 3 XP_016857542.1
XM_017002054.1 2323 UTR 3 XP_016857543.1

View Full Product Details