Product Details

SNP ID
rs112980322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46923312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGAGTCAGCCTGCACCCGCCAC[A/G]CACGGGCAACACTCACCAGTCAGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC12 PubMed Links

Gene Details

Gene
CCDC12
Gene Name
coiled-coil domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277074.1 Intron NP_001264003.1
NM_144716.5 Intron NP_653317.2
XM_011533390.2 Intron XP_011531692.1
XM_011533391.2 Intron XP_011531693.1
XM_011533392.2 Intron XP_011531694.1
XM_011533393.2 Intron XP_011531695.1
XM_011533394.2 Intron XP_011531696.1
XM_011533396.2 Intron XP_011531698.1

View Full Product Details