Product Details

SNP ID
rs57195580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187199798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACGGCGAGCGTGGCGGCCAGTAC[C/T]GCATCCACGTGGCCAGCCGCCAGGA
Phenotype
MIM: 609137
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929106 PubMed Links

Gene Details

Gene
LOC101929106
Gene Name
uncharacterized LOC101929106
There are no transcripts associated with this gene.

Gene
RTP1
Gene Name
receptor transporter protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153708.2 550 Missense Mutation CGC,TGC R174C NP_714919.2

View Full Product Details