Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039724.3 | 2610 | UTR 3 | NP_001034813.2 | ||
NM_001171631.1 | 2610 | UTR 3 | NP_001165102.1 | ||
NM_001171632.1 | 2610 | UTR 3 | NP_001165103.1 | ||
NM_052946.3 | 2610 | UTR 3 | NP_443178.2 | ||
XM_005246270.1 | 2610 | Intron | XP_005246327.1 | ||
XM_006712233.3 | 2610 | Intron | XP_006712296.1 | ||
XM_011510540.2 | 2610 | Intron | XP_011508842.1 | ||
XM_011510543.2 | 2610 | Intron | XP_011508845.1 | ||
XM_017003278.1 | 2610 | Intron | XP_016858767.1 | ||
XM_017003279.1 | 2610 | Intron | XP_016858768.1 | ||
XM_017003280.1 | 2610 | Intron | XP_016858769.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020675.3 | 2610 | Intron | NP_065726.1 | ||
XM_011511516.2 | 2610 | UTR 3 | XP_011509818.1 |