Product Details

SNP ID
rs61745549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34739556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTAACTAAGCCATATGTGATCA[C/T]GTTATTGGAGGATGGAAAAGAGCCC
Phenotype
MIM: 606741
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF181 PubMed Links

Gene Details

Gene
ZNF181
Gene Name
zinc finger protein 181
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029997.3 395 Missense Mutation ACG,ATG T55M NP_001025168.2
NM_001145665.1 395 Missense Mutation ACG,ATG T54M NP_001139137.1
XM_005258849.2 395 Missense Mutation ACG,ATG T55M XP_005258906.1
XM_005258850.2 395 Missense Mutation ACG,ATG T54M XP_005258907.1
XM_005258851.3 395 Missense Mutation ACG,ATG T55M XP_005258908.1
XM_006723183.3 395 Missense Mutation ACG,ATG T55M XP_006723246.1
XM_006723184.3 395 Missense Mutation ACG,ATG T54M XP_006723247.1
XM_017026739.1 395 Missense Mutation ACG,ATG T54M XP_016882228.1
XM_017026740.1 395 Missense Mutation ACG,ATG T73M XP_016882229.1

View Full Product Details