Product Details

SNP ID
rs61743324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52065389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGTAAACCAAGTTTGACCTTTC[A/G]ATAAAAGCTTTACCACATTCATTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF841 PubMed Links

Gene Details

Gene
ZNF841
Gene Name
zinc finger protein 841
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136499.1 4548 Silent Mutation ATC,ATT I831I NP_001129971.1
NM_001321349.1 4548 Silent Mutation ATC,ATT I715I NP_001308278.1
XM_011526833.2 4548 Intron XP_011525135.1
XM_017026652.1 4548 Intron XP_016882141.1
XM_017026653.1 4548 Silent Mutation ATC,ATT I832I XP_016882142.1
XM_017026654.1 4548 Silent Mutation ATC,ATT I832I XP_016882143.1
XM_017026655.1 4548 Intron XP_016882144.1
XM_017026656.1 4548 Silent Mutation ATC,ATT I831I XP_016882145.1
XM_017026657.1 4548 Intron XP_016882146.1
XM_017026658.1 4548 Intron XP_016882147.1
XM_017026659.1 4548 Intron XP_016882148.1
XM_017026660.1 4548 Intron XP_016882149.1
XM_017026661.1 4548 Intron XP_016882150.1
XM_017026662.1 4548 Intron XP_016882151.1
XM_017026663.1 4548 Intron XP_016882152.1

View Full Product Details