Product Details

SNP ID
rs368307055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55602799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCGGAGGCCATGGGGGTACCCCT[G/T]TGTGCACCAGATCCAGGGTCTGAAC
Phenotype
MIM: 609133
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FIZ1 PubMed Links

Gene Details

Gene
FIZ1
Gene Name
FLT3 interacting zinc finger 1
There are no transcripts associated with this gene.

Gene
ZNF524
Gene Name
zinc finger protein 524
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153219.3 1035 Silent Mutation CTG,CTT L229L NP_694951.1
XM_011526487.2 1035 Silent Mutation CTG,CTT L335L XP_011524789.1

View Full Product Details