Product Details

SNP ID
rs9902379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76737283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGACACTGGGAAAGGCCTTGCCG[C/T]AGAACAGCACGGACGGGCTCCGCAG
Phenotype
MIM: 615262 MIM: 600813
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
METTL23 PubMed Links

Gene Details

Gene
METTL23
Gene Name
methyltransferase like 23
There are no transcripts associated with this gene.

Gene
MFSD11
Gene Name
major facilitator superfamily domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242532.1 129 Intron NP_001229461.1
NM_001242533.1 129 Intron NP_001229462.1
NM_001242534.1 129 Intron NP_001229463.1
NM_001242535.1 129 Intron NP_001229464.1
NM_001242536.1 129 Intron NP_001229465.1
NM_001242537.1 129 Intron NP_001229466.1
NM_024311.3 129 Intron NP_077287.1
XM_011525236.2 129 Intron XP_011523538.1
XM_011525237.2 129 Intron XP_011523539.2
XM_011525238.2 129 Intron XP_011523540.2
XM_011525239.2 129 Intron XP_011523541.2
XM_011525240.2 129 Intron XP_011523542.2
XM_011525241.2 129 Intron XP_011523543.2
XM_011525242.1 129 Intron XP_011523544.1
XM_011525244.1 129 Intron XP_011523546.1
XM_011525247.2 129 Intron XP_011523549.2
XM_017025065.1 129 Intron XP_016880554.1
XM_017025066.1 129 Intron XP_016880555.1
XM_017025067.1 129 Intron XP_016880556.1
XM_017025068.1 129 Intron XP_016880557.1
XM_017025069.1 129 Intron XP_016880558.1
XM_017025070.1 129 Intron XP_016880559.1
XM_017025071.1 129 Intron XP_016880560.1
Gene
MIR636
Gene Name
microRNA 636
There are no transcripts associated with this gene.

Gene
SRSF2
Gene Name
serine and arginine rich splicing factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195427.1 129 UTR 5 NP_001182356.1
NM_003016.4 129 UTR 5 NP_003007.2
XM_017024942.1 129 UTR 5 XP_016880431.1

View Full Product Details