Product Details

SNP ID
rs79163931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23837178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCGCAGCCTCCTCTCTTCTGCAG[A/G]AGTGAAGGCAGATGGGGGTTACAGC
Phenotype
MIM: 176970
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKCB PubMed Links

Gene Details

Gene
PRKCB
Gene Name
protein kinase C beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002738.6 Intron NP_002729.2
NM_212535.2 Intron NP_997700.1

View Full Product Details