Product Details

SNP ID
rs150406105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118531918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCAAGTCCTTCTGGGTTCCAAAG[A/C]CCCCTCTCTCCCCTGTCTGCACTTC
Phenotype
MIM: 159555 MIM: 613934
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KMT2A PubMed Links

Gene Details

Gene
KMT2A
Gene Name
lysine methyltransferase 2A
There are no transcripts associated with this gene.

Gene
LOC101929089
Gene Name
uncharacterized LOC101929089
There are no transcripts associated with this gene.

Gene
TMEM25
Gene Name
transmembrane protein 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144034.1 Intron NP_001137506.1
NM_001144035.1 Intron NP_001137507.1
NM_001144036.1 Intron NP_001137508.1
NM_001144037.1 Intron NP_001137509.1
NM_001144038.1 Intron NP_001137510.1
NM_001318755.1 Intron NP_001305684.1
NM_001318757.1 Intron NP_001305686.1
NM_032780.3 Intron NP_116169.2
XM_005271703.1 Intron XP_005271760.1
XM_006718926.2 Intron XP_006718989.1
XM_006718927.2 Intron XP_006718990.1
XM_011543037.2 Intron XP_011541339.1
XM_011543038.2 Intron XP_011541340.1
XM_017018422.1 Intron XP_016873911.1
XM_017018423.1 Intron XP_016873912.1
XM_017018424.1 Intron XP_016873913.1
XM_017018425.1 Intron XP_016873914.1
XM_017018426.1 Intron XP_016873915.1
XM_017018427.1 Intron XP_016873916.1
XM_017018428.1 Intron XP_016873917.1
XM_017018429.1 Intron XP_016873918.1
XM_017018430.1 Intron XP_016873919.1
XM_017018431.1 Intron XP_016873920.1
XM_017018432.1 Intron XP_016873921.1
XM_017018433.1 Intron XP_016873922.1
Gene
TTC36
Gene Name
tetratricopeptide repeat domain 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080441.1 Intron NP_001073910.1

View Full Product Details