Product Details

SNP ID
rs13338974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28101742 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCAGCCCCCAGGGGCCTGTCCCG[A/G]GTCCCATCCACTTTCTGTCACACTC
Phenotype
MIM: 608411
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XPO6 PubMed Links

Gene Details

Gene
XPO6
Gene Name
exportin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270940.1 Intron NP_001257869.1
NM_015171.3 Intron NP_055986.1
XM_005255197.3 Intron XP_005255254.1
XM_005255198.1 Intron XP_005255255.1
XM_005255199.2 Intron XP_005255256.1
XM_011545772.2 Intron XP_011544074.1

View Full Product Details