Product Details

SNP ID
rs145897859
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:48728263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAAGTAATGATAAAGTGTCCAC[A/G]TGTCCACCACTCAGTGGAGCAGGAT
Phenotype
MIM: 616476
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGBL4 PubMed Links

Gene Details

Gene
AGBL4
Gene Name
ATP/GTP binding protein like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323573.1 Intron NP_001310502.1
NM_001323574.1 Intron NP_001310503.1
NM_001323575.1 Intron NP_001310504.1
NM_032785.3 Intron NP_116174.3
XM_011542308.2 Intron XP_011540610.1
XM_011542310.2 Intron XP_011540612.1
XM_017002595.1 Intron XP_016858084.1
XM_017002596.1 Intron XP_016858085.1
XM_017002597.1 Intron XP_016858086.1
XM_017002598.1 Intron XP_016858087.1
Gene
BEND5
Gene Name
BEN domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302082.1 Intron NP_001289011.1
NM_024603.3 Intron NP_078879.2
XM_011542141.2 Intron XP_011540443.1
XM_011542142.2 Intron XP_011540444.1
XM_017002331.1 Intron XP_016857820.1
XM_017002332.1 Intron XP_016857821.1
XM_017002333.1 Intron XP_016857822.1
XM_017002334.1 Intron XP_016857823.1

View Full Product Details