Product Details

SNP ID
rs116527400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30287116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGGGACCCAGTGGGAAGAGGA[A/G]GCCCTGCCCAAGTCCAAGTGTCAGG
Phenotype
MIM: 617034 MIM: 610020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GATSL3 PubMed Links

Gene Details

Gene
GATSL3
Gene Name
GATS protein like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037666.2 Intron NP_001032755.1
Gene
TBC1D10A
Gene Name
TBC1 domain family member 10A
There are no transcripts associated with this gene.

View Full Product Details