Product Details

SNP ID
hCV25962547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:48085396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGGGAAGCCTGGGAGGGGAAG[A/G]TGCTCTTGCTGTAAATGGCTGTTAC
Phenotype
MIM: 607041
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCC12 PubMed Links

Gene Details

Gene
ABCC12
Gene Name
ATP binding cassette subfamily C member 12
There are no transcripts associated with this gene.

View Full Product Details