Product Details

SNP ID
rs139904707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66686158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGGACACTGTGCTGAGTTGGCC[A/G]CAGTGGACAGCAGGGAAGTGTAAGA
Phenotype
MIM: 604985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM4B PubMed Links

Gene Details

Gene
RBM4B
Gene Name
RNA binding motif protein 4B
There are no transcripts associated with this gene.

Gene
SPTBN2
Gene Name
spectrin beta, non-erythrocytic 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006946.2 Intron NP_008877.1
XM_005274192.4 Intron XP_005274249.1
XM_005274193.3 Intron XP_005274250.1
XM_006718669.3 Intron XP_006718732.1
XM_006718671.3 Intron XP_006718734.1
XM_011545216.2 Intron XP_011543518.1
XM_011545217.2 Intron XP_011543519.1
XM_017018174.1 Intron XP_016873663.1
XM_017018175.1 Intron XP_016873664.1
XM_017018176.1 Intron XP_016873665.1
XM_017018177.1 Intron XP_016873666.1
XM_017018178.1 Intron XP_016873667.1

View Full Product Details