Product Details

SNP ID
rs45513592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49646556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACATGGCCACGGACAGCTTCCTC[G/T]CAGGGCTGGTGAGTGTCCTGGATCC
Phenotype
MIM: 165090
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RRAS PubMed Links

Gene Details

Gene
RRAS
Gene Name
related RAS viral (r-ras) oncogene homolog
There are no transcripts associated with this gene.

Gene
SCAF1
Gene Name
SR-related CTD associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021228.2 143 Missense Mutation GCA,TCA A98S NP_067051.2
XM_005259122.4 143 Missense Mutation GCA,TCA A98S XP_005259179.1
XM_011527194.2 143 Missense Mutation GCA,TCA A101S XP_011525496.1
XM_017027083.1 143 Missense Mutation GCA,TCA A8S XP_016882572.1

View Full Product Details