Product Details

SNP ID
rs114843375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49818330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGCTGCAGTGGTGGTGGCGGGT[A/G]CCGCACGGGGTATGGTCCCCGGGTC
Phenotype
MIM: 610622 MIM: 610197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FUZ PubMed Links

Gene Details

Gene
FUZ
Gene Name
fuzzy planar cell polarity protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171937.1 52 Intron NP_001165408.1
NM_025129.4 52 Intron NP_079405.2
XM_006723399.3 52 Intron XP_006723462.1
XM_011527339.1 52 Intron XP_011525641.1
XM_011527340.1 52 Intron XP_011525642.1
XM_011527341.2 52 Intron XP_011525643.1
XM_011527342.1 52 Intron XP_011525644.1
XM_011527343.1 52 Intron XP_011525645.1
XM_011527345.1 52 Intron XP_011525647.1
XM_011527346.1 52 Intron XP_011525648.1
XM_011527347.1 52 Intron XP_011525649.1
XM_017027319.1 52 Intron XP_016882808.1
XM_017027320.1 52 Intron XP_016882809.1
XM_017027321.1 52 Intron XP_016882810.1
XM_017027322.1 52 Intron XP_016882811.1
XM_017027323.1 52 Intron XP_016882812.1
Gene
MED25
Gene Name
mediator complex subunit 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030973.3 52 UTR 5 NP_112235.2
XM_011527353.1 52 UTR 5 XP_011525655.1

View Full Product Details