Product Details

SNP ID
rs35662948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54161144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGCACTTACAGGAAGATGCTGTA[A/G]AGCAGGGGAACGCTGGAGATGGCCA
Phenotype
MIM: 604910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT3 PubMed Links

Gene Details

Gene
CNOT3
Gene Name
CCR4-NOT transcription complex subunit 3
There are no transcripts associated with this gene.

Gene
LENG1
Gene Name
leukocyte receptor cluster member 1
There are no transcripts associated with this gene.

Gene
TMC4
Gene Name
transmembrane channel like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145303.2 1439 Silent Mutation CTC,CTT L607L NP_001138775.2
NM_144686.3 1439 Silent Mutation CTC,CTT L601L NP_653287.2
XM_011526486.2 1439 Silent Mutation CTC,CTT L447L XP_011524788.1

View Full Product Details