Product Details

SNP ID
rs12373105
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.16:48083748 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
TGACTTCTGCTGCTAGTAACATCGC[A/T]AATGCAGAATCTGGCTTCTCTGCAA
Phenotype
MIM: 607041
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCC12 PubMed Links

Gene Details

Gene
ABCC12
Gene Name
ATP binding cassette subfamily C member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033226.2 4393 Missense Mutation TTA,TTT L1349F NP_150229.2

View Full Product Details