Product Details

SNP ID
rs896030529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:98384019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCACTGGAATCACCTAAGACAGC[C/T]GCTCAATAGATTCTGGCTCCCTCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PYROXD2 PubMed Links

Gene Details

Gene
PYROXD2
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032709.2 Intron NP_116098.2

View Full Product Details