Product Details

SNP ID
rs185436240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:100704482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATACCAATTCCCAACAAAGATA[A/G]GTTTCTTCCCTCAAATGAAAGTATA
Phenotype
MIM: 604679
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR7705 PubMed Links

Gene Details

Gene
MIR7705
Gene Name
microRNA 7705
There are no transcripts associated with this gene.

Gene
PABPC1
Gene Name
poly(A) binding protein cytoplasmic 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002568.3 Intron NP_002559.2
XM_005250861.2 Intron XP_005250918.1

View Full Product Details