Product Details

SNP ID
rs34957169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27911619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACATGTCCCAAATGCCTTTCGT[C/T]GACCTTCAGCAGACTGGATTCTCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2B2 PubMed Links

Gene Details

Gene
OR2B2
Gene Name
olfactory receptor family 2 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033057.2 778 Missense Mutation CAA,CGA Q234R NP_149046.2

View Full Product Details