Product Details

SNP ID
rs57864418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:237088587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTTTTAATTGATTATTCTTCTT[C/T]GTGGTGGCGTAAATTAGGAATAATG
Phenotype
MIM: 616011
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPS8 PubMed Links

Gene Details

Gene
COPS8
Gene Name
COP9 signalosome subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006710.4 Intron NP_006701.1
NM_198189.2 Intron NP_937832.1

View Full Product Details