Product Details

SNP ID
rs114682066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44684071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCATGCTGTGAAATTATGGAA[A/G]GGTCTGGGAGCCACAATAGAGTATG
Phenotype
MIM: 616934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC43 PubMed Links

Gene Details

Gene
CCDC43
Gene Name
coiled-coil domain containing 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099225.1 Intron NP_001092695.1
NM_144609.2 Intron NP_653210.2
Gene
MEIOC
Gene Name
meiosis specific with coiled-coil domain
There are no transcripts associated with this gene.

View Full Product Details