Product Details

SNP ID
rs35065609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23097704 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAATGCCACTATCCTTCCCATCT[C/G]TCTTACCCTCAGTACCACACAATAC
Phenotype
MIM: 604562
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACIN1 PubMed Links

Gene Details

Gene
ACIN1
Gene Name
apoptotic chromatin condensation inducer 1
There are no transcripts associated with this gene.

Gene
C14orf119
Gene Name
chromosome 14 open reading frame 119
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017924.3 193 Missense Mutation CTC,GTC L16V NP_060394.1
XM_017021390.1 193 Missense Mutation CTC,GTC L16V XP_016876879.1
Gene
LOC100128908
Gene Name
leishmanolysin homolog
There are no transcripts associated with this gene.

View Full Product Details