Product Details

SNP ID
rs36084440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44172222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCGAGCCCAGTTTGACTGTGTG[A/C]GGCTGCTGCTGACCTTCGGAGCCAA
Phenotype
MIM: 615056
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ASB16 PubMed Links

Gene Details

Gene
ASB16
Gene Name
ankyrin repeat and SOCS box containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080863.4 562 Silent Mutation AGG,CGG R160R NP_543139.4
Gene
ASB16-AS1
Gene Name
ASB16 antisense RNA 1
There are no transcripts associated with this gene.

Gene
C17orf53
Gene Name
chromosome 17 open reading frame 53
There are no transcripts associated with this gene.

View Full Product Details