Product Details

SNP ID
rs112328838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63590052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGCCGCGGCTGAGAGACAGCC[A/T]GCCCTGTCCGTCCCAGGGGCCTCGC
Phenotype
MIM: 607451
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GMEB2 PubMed Links

Gene Details

Gene
GMEB2
Gene Name
glucocorticoid modulatory element binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012384.4 1485 UTR 3 NP_036516.1
XM_005260202.4 1485 UTR 3 XP_005260259.1
XM_006723776.2 1485 UTR 3 XP_006723839.1
XM_011528779.2 1485 UTR 3 XP_011527081.1

View Full Product Details